Learn More
Optimize Technologies OPTI-LYNX™ Direct-Connect Union Kit (Includes: UPA, holder cap/tube, bayonet connector, sealing element & fitting)
SDP
Catalog No. 50113440
Encompass_Preferred
If you are already using our OPTI-LYNX™ Guard Column/Pre-Column Filter Kits and want to use OPTI-LYNX™ at the bottom of the column as well, upgrade with this kit. It includes everything you need to equip one end of any analytical column with straight-through quick-connect operation. Can be used alone or in conjunction with other OPTI-LYNX™ products.
Catalog No. 50-113-440 Supplier Optimize Technologies Supplier No. 1102226AB
If you are already using our OPTI-LYNX™ Guard Column/Pre-Column Filter Kits and want to use OPTI-LYNX™ at the bottom of the column as well, upgrade with this kit. It includes everything you need to equip one end of any analytical column with straight-through quick-connect operation. Can be used alone or in conjunction with other OPTI-LYNX™ products.