Learn More
Optimize Technologies OPTI-LYNX™ Direct-Connect Guard Column Holder Kit (OPTI-LYNX™ guard cartridges ordered separately)
SDP
Catalog No. 50113441
Encompass_Preferred
The OPTI-LYNX™ Guard Column is a high-performance, cartridge-based guard system, suitable for use with any 10-32 analytical column due to its auto-adjusting ZDV port adapter. OPTI-LYNX™ combines effective column protection with the convenience of quick-connect operation. Some cartridge-based guard column products offer convenience with a price: higher internal volumes. At Optimize Technologies, we refuse to compromise performance for anything. OPTI-LYNX™ utilizes low dispersion component interfaces to keep extra column volume to a minimum: connection paths have a total internal volume of less than 2uL. You get quick-connect convenience and low dispersion, all in one package.
Catalog No. 50-113-441 Supplier Optimize Technologies Supplier No. 1102232AA
The OPTI-LYNX™ Guard Column is a high-performance, cartridge-based guard system, suitable for use with any 10-32 analytical column due to its auto-adjusting ZDV port adapter. OPTI-LYNX™ combines effective column protection with the convenience of quick-connect operation. Some cartridge-based guard column products offer convenience with a price: higher internal volumes. At Optimize Technologies, we refuse to compromise performance for anything. OPTI-LYNX™ utilizes low dispersion component interfaces to keep extra column volume to a minimum: connection paths have a total internal volume of less than 2uL. You get quick-connect convenience and low dispersion, all in one package.