Learn More
Optimize Technologies OPTI-LYNX™ Holder Cap
SDP
Catalog No. 50113460
Encompass_Preferred
All OPTI-LYNX™ hardware can be purchased in kits or separately as needed for your application. OPTI-LYNX™ hardware is precision machined to maintain the highest quality and strictest tolerances. Switching between modes of pre-column protection is as easy as opening OPTI-LYNX™ hardware. Simply select the correct holder tube for the OPTI-LYNX™ insert you wish to use (a guard column, pre-column filter, or straight through connection without protection) thread it into the holder cap, pop in your insert, you're ready to go.
Catalog No. 50-113-460 Supplier Optimize Technologies Supplier No. 1102333AA
All OPTI-LYNX™ hardware can be purchased in kits or separately as needed for your application. OPTI-LYNX™ hardware is precision machined to maintain the highest quality and strictest tolerances. Switching between modes of pre-column protection is as easy as opening OPTI-LYNX™ hardware. Simply select the correct holder tube for the OPTI-LYNX™ insert you wish to use (a guard column, pre-column filter, or straight through connection without protection) thread it into the holder cap, pop in your insert, you're ready to go.