Learn More
Optimize Technologies OPTI-LYNX™ Sealing Element
SDP
Catalog No. 50113453
Encompass_Preferred
All OPTI-LYNX™ hardware can be purchased in kits or separately as needed for your application. OPTI-LYNX™ hardware is precision machined to maintain the highest quality and strictest tolerances. Use an OPTI-LYNX™ direct-connect in-line kit & an extra bayonet connector to switch quickly and easily between multiple detectors or flow paths. Simply terminate the column outlet line with a holder cap/tube and use extra bayonet connectors for each detector inlet line. This product provides a straight-through fluid connection when filters and packed beds are not required.
Catalog No. 50-113-453 Supplier Optimize Technologies Supplier No. 1102307AB
All OPTI-LYNX™ hardware can be purchased in kits or separately as needed for your application. OPTI-LYNX™ hardware is precision machined to maintain the highest quality and strictest tolerances. Use an OPTI-LYNX™ direct-connect in-line kit & an extra bayonet connector to switch quickly and easily between multiple detectors or flow paths. Simply terminate the column outlet line with a holder cap/tube and use extra bayonet connectors for each detector inlet line. This product provides a straight-through fluid connection when filters and packed beds are not required.