Learn More
Optimize Technologies OPTI-LYNX™ Universal Port Adapter (UPA) - Bayonet Connector w/ Auto-adjusting 10-32 Male Connector
SDP
Catalog No. 50113451
Encompass_Preferred
All OPTI-LYNX™ hardware can be purchased in kits or separately as needed for your application. OPTI-LYNX™ hardware is precision machined to maintain the highest quality and strictest tolerances. The Universal Port Adapter with Auto-adjusting 10-32 Male Connector provides a zero-dead-volume connection regardless of the mating port depth. It also adapts any 10-32 column or detector port to OPTI-LYNX™ hardware. Set up a simple column-switching rig without the need for expensive high-pressure valves - simply install OPTI-LYNX™ universal port adapters on columns you're getting ready to use, and switch columns quickly and easily (pressure should be allowed to dissipate before column is disconnected).
Catalog No. 50-113-451 Supplier Optimize Technologies Supplier No. 1102290AA
All OPTI-LYNX™ hardware can be purchased in kits or separately as needed for your application. OPTI-LYNX™ hardware is precision machined to maintain the highest quality and strictest tolerances. The Universal Port Adapter with Auto-adjusting 10-32 Male Connector provides a zero-dead-volume connection regardless of the mating port depth. It also adapts any 10-32 column or detector port to OPTI-LYNX™ hardware. Set up a simple column-switching rig without the need for expensive high-pressure valves - simply install OPTI-LYNX™ universal port adapters on columns you're getting ready to use, and switch columns quickly and easily (pressure should be allowed to dissipate before column is disconnected).