Learn More
Optimize Technologies OPTI-MAX Outlet Cartridge Check Valve, 1/8" Ceramic Ball & Seat, SS Cartridge, Waters 100 µL Head [M45/45G, M501, 510, 590, 600/600E, 610, 6K/6KA] & 225 µL Head (#3) 515, 1515, 1525
SDP
Catalog No. 50114252
Encompass_Preferred
Opti-Max™ Cartridge Check Valves are designed to offer maximum performance and ease of use. From the sharp, consistent threads on OPTI-MAX™ custom housings, to the exacting standards used to match ball/seat sets in OPTI-MAX™ cartridges, every aspect of design and manufacturing reflects an unavoidable fact: when it comes to reliable check valve operation, quality and precision make a big difference. OPTI-MAX™ check valves offer many advantages over OEM check valve systems, and are better designed, better made and more reliable than any other after-market check valve product. Additionally, OPTI-MAX™ Free-Turn™ housings can be removed with the tubing still attached, accelerating check valve maintenance while reducing fitting wear. This item is equivalent to OEM part numbers WAT033679 and 700000253.
Catalog No. 50-114-252 Supplier Optimize Technologies Supplier No. 395601256
Opti-Max™ Cartridge Check Valves are designed to offer maximum performance and ease of use. From the sharp, consistent threads on OPTI-MAX™ custom housings, to the exacting standards used to match ball/seat sets in OPTI-MAX™ cartridges, every aspect of design and manufacturing reflects an unavoidable fact: when it comes to reliable check valve operation, quality and precision make a big difference. OPTI-MAX™ check valves offer many advantages over OEM check valve systems, and are better designed, better made and more reliable than any other after-market check valve product. Additionally, OPTI-MAX™ Free-Turn™ housings can be removed with the tubing still attached, accelerating check valve maintenance while reducing fitting wear. This item is equivalent to OEM part numbers WAT033679 and 700000253.