Learn More
Optimize Technologies OPTI-MAX Outlet Cartridge Check Valve, 1/8" Ceramic Ball & Seat, PEEK Cartridge, Waters 100 µL Head [M45/45G, M501, 510, 590, 600/600E, 610, 6K/6KA] & 225 µL Head (#3)
SDP
Catalog No. 50114260
Encompass_Preferred
Opti-Max™ Cartridge Check Valves are designed to offer maximum performance and ease of use. From the sharp, consistent threads on OPTI-MAX™ custom housings, to the exacting standards used to match ball/seat sets in OPTI-MAX™ cartridges, every aspect of design and manufacturing reflects an unavoidable fact: when it comes to reliable check valve operation, quality and precision make a big difference. OPTI-MAX™ check valves offer many advantages over OEM check valve systems, and are better designed, better made and more reliable than any other after-market check valve product. Additionally, OPTI-MAX™ Free-Turn™ housings can be removed with the tubing still attached, accelerating check valve maintenance while reducing fitting wear.
Catalog No. 50-114-260 Supplier Optimize Technologies Supplier No. 395801256
Opti-Max™ Cartridge Check Valves are designed to offer maximum performance and ease of use. From the sharp, consistent threads on OPTI-MAX™ custom housings, to the exacting standards used to match ball/seat sets in OPTI-MAX™ cartridges, every aspect of design and manufacturing reflects an unavoidable fact: when it comes to reliable check valve operation, quality and precision make a big difference. OPTI-MAX™ check valves offer many advantages over OEM check valve systems, and are better designed, better made and more reliable than any other after-market check valve product. Additionally, OPTI-MAX™ Free-Turn™ housings can be removed with the tubing still attached, accelerating check valve maintenance while reducing fitting wear.