Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Gibco™ Opti-MEM™ I Reduced Serum Medium
GREENER_CHOICE

Catalog No. 31985062
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 mL
500 mL
10 x 500 mL
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
31-985-062 100 mL
31-985-070 500 mL
31-985-088 10 x 500 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Catalog No. 31-985-062 Supplier Gibco™ Supplier No. 31985062
Only null left
Add to Cart
Edge
Add to Cart
This item is not returnable. View return policy

Opti-MEM™ I Reduced-Serum Medium is an improved Minimal Essential Medium (MEM) that allows for a reduction of Fetal Bovine Serum supplementation by at least 50% with no change in growth rate or morphology.

Opti-MEM™ I Reduced-Serum Medium is an improved Minimal Essential Medium (MEM) that allows for a reduction of Fetal Bovine Serum supplementation by at least 50% with no change in growth rate or morphology. Opti-MEM™ I medium is also recommended for use with cationic lipid transfection reagents, such as Lipofectamine™ reagent. Opti-MEM™ I medium can be used with a variety of suspension and adherent mammalian cells, including Sp2, AE-1, CHO, BHK-21, HEK, and primary fibroblasts. We offer a variety of Opti-MEM™ I modifications for a range of cell culture applications.

This Opti-MEM™ I is modified as follows:

With: L-glutamine, Phenol Red

The complete formulation is confidential. For more information, please contact Technical Services.

Using Opti-MEM™ I Medium

Opti-MEM™ I Reduced-Serum Medium is a unique medium that contains insulin, transferrin, hypoxanthine, thymidine, and trace elements. These additional components allow for a reduction in serum supplementation by at least 50%. Opti-MEM™ I medium uses a sodium bicarbonate buffer system (2.4 g/L), and therefore requires a 5–10% CO2 environment to maintain physiological pH.

Order Info

Shipping Condition: Room Temperature

TRUSTED_SUSTAINABILITY

Specifications

Cell Line Sp2, AE-1, CHO, BHK-21, and HEK
Cell Type Primary Fibroblasts
Classification Human Origin
Concentration 1 X
Form Liquid
Product Type Opti-MEM I
Sterility Sterile-filtered
Sterilization Method Sterile-filtered
With Additives Glutamine, Phenol Red
Manufacturing Quality cGMP-compliant under the ISO 13485 standard
Product Line Gibco, Opti-MEM
Quantity 100 mL
Shipping Condition Room Temperature
Content And Storage Store at 2–8°C and protect from light.
Show More Show Less

For Research Use or Further Manufacturing. Not for diagnostic use or direct administration into humans or animals.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.