Learn More
ELITechGroup OPTIMOLE™ Osmolality Standard, 100 mmol/kg, 60x0.4mL
SDP
Catalog No. NC9041262
Encompass
Wescor Osmolality Standard, 100mmol/kg, Chemical Name: Sodium chloride solution, Liquid, Odorless, Clear, Freezing Point: 0 deg.C, b.p.: 100 deg.C, Solubility in water, Max Storage Temperature: 31 deg.C
Catalog No. NC9041262 Supplier ELITechGroup Supplier No. OA010
Wescor Osmolality Standard, 100mmol/kg, Chemical Name: Sodium chloride solution, Liquid, Odorless, Clear, Freezing Point: 0 deg.C, b.p.: 100 deg.C, Solubility in water, Max Storage Temperature: 31 deg.C