Learn More
Optimize Technologies OPTI-SEAL UHMW-PE Piston Seal (Hastelloy C Spring), Dionex/All Analytical
SDP
Catalog No. 50113704
Encompass_Preferred
Optimize Piston Seals are the longest lasting seals on the market. Machined from either of two proprietary polymer blends, UHMW-PE or ITB, Optimize Seals provide the ultimate in high pressure sealing capacity. For mostly aqueous buffered mobile phases, Opti-Seal™ UHMW-polyethylene is the wisest choice. Specially designed to be more resistant to abrasion and wear than standard polyethylene seals, they can last up to seven times longer under mostly aqueous conditions. Despite the UHMW-PE's durability, it has a coefficient of friction approaching that of PTFE. The homogenous polymer nature leads to less shedding, but when it does occur, a larger, more fibrous particle is produced which is easily caught by a filter frit.
Catalog No. 50-113-704 Supplier Optimize Technologies Supplier No. 253600597
Optimize Piston Seals are the longest lasting seals on the market. Machined from either of two proprietary polymer blends, UHMW-PE or ITB, Optimize Seals provide the ultimate in high pressure sealing capacity. For mostly aqueous buffered mobile phases, Opti-Seal™ UHMW-polyethylene is the wisest choice. Specially designed to be more resistant to abrasion and wear than standard polyethylene seals, they can last up to seven times longer under mostly aqueous conditions. Despite the UHMW-PE's durability, it has a coefficient of friction approaching that of PTFE. The homogenous polymer nature leads to less shedding, but when it does occur, a larger, more fibrous particle is produced which is easily caught by a filter frit.