Learn More
Optimize Technologies OPTI-SEAL UHMW-PE Wash Seal HP/Agilent 1050, 1100, 1200, 10pk
SDP
Catalog No. 50113766
Encompass_Preferred
Optimize Piston Seals are the longest lasting seals on the market. Machined from either of two proprietary polymer blends, UHMW-PE or ITB, Optimize Seals provide the ultimate in high pressure sealing capacity. For mostly aqueous buffered mobile phases, Opti-Seal™ UHMW-polyethylene is the wisest choice. Specially designed to be more resistant to abrasion and wear than standard polyethylene seals, they can last up to seven times longer under mostly aqueous conditions. Despite the UHMW-PE's durability, it has a coefficient of friction approaching that of PTFE. The homogenous polymer nature leads to less shedding, but when it does occur, a larger, more fibrous particle is produced which is easily caught by a filter frit. This item is equivalent to OEM part number 0905-1175.
Catalog No. 50-113-766 Supplier Optimize Technologies Supplier No. 273602365
Optimize Piston Seals are the longest lasting seals on the market. Machined from either of two proprietary polymer blends, UHMW-PE or ITB, Optimize Seals provide the ultimate in high pressure sealing capacity. For mostly aqueous buffered mobile phases, Opti-Seal™ UHMW-polyethylene is the wisest choice. Specially designed to be more resistant to abrasion and wear than standard polyethylene seals, they can last up to seven times longer under mostly aqueous conditions. Despite the UHMW-PE's durability, it has a coefficient of friction approaching that of PTFE. The homogenous polymer nature leads to less shedding, but when it does occur, a larger, more fibrous particle is produced which is easily caught by a filter frit. This item is equivalent to OEM part number 0905-1175.