Learn More
Optimize Technologies OPTI-SOLV Analytical Solvent Reservoir Filter for 1/8" OD Tubing, 5pk
SDP
Catalog No. 50113239
Encompass_Preferred
A solid PTFE body keeps the OPTI-SOLV™ filter at the base of the bottle where a bottom-mounted titanium frit provides unrestricted solvent access. As always, Optimize makes installation as simple as possible - no tools or fittings required and includes 5 feet of pre-installed PTFE tubing. OPTI-SOLV™ Reservoir Filters employ a unique conical design inside the filter housing to prevent air bubbles from getting trapped and disturbing your analysis. While the availability of HPLC-grade solvents diminishes the need for pre-filtering, it is still imperative that a filtration device be installed in the reservoir. Placing a filter in the reservoir is an economical way to filter particles that may result from buffer salt precipitation, airborne dust, improperly cleaned glassware, or microbial contamination; none of which are remedied by the use of HPLC-grade solvents.
Catalog No. 50-113-239 Supplier Optimize Technologies Supplier No. 100400053
A solid PTFE body keeps the OPTI-SOLV™ filter at the base of the bottle where a bottom-mounted titanium frit provides unrestricted solvent access. As always, Optimize makes installation as simple as possible - no tools or fittings required and includes 5 feet of pre-installed PTFE tubing. OPTI-SOLV™ Reservoir Filters employ a unique conical design inside the filter housing to prevent air bubbles from getting trapped and disturbing your analysis. While the availability of HPLC-grade solvents diminishes the need for pre-filtering, it is still imperative that a filtration device be installed in the reservoir. Placing a filter in the reservoir is an economical way to filter particles that may result from buffer salt precipitation, airborne dust, improperly cleaned glassware, or microbial contamination; none of which are remedied by the use of HPLC-grade solvents.