Learn More
Optimize Technologies OPTI-SOLV In-Line Kit, 2.0µm, Includes Hand-Tight PEEK Holder, Fittings & 2 Filter Elements
SDP
Supplier:  Optimize Technologies 100400391
Encompass_Preferred
The user-friendly Hand-Tight OPTI-SOLV™ In-Line Filter system is well suited for a number of applications, such as high temperature, high-pressure environments where quick changes are imperative. The unique swivel design allows frits to be changed without removing the connecting tubing. Just plumb it once and leave it. The frictional forces generated at the piston/seal interface inevitably lead to particulate shedding. Placement of a filtering element between the pump and injector/ auto sampler removes these particulates, preventing problems with downstream components and costly downtime. Key Features include: a holder, fittings and 2 filter elements, the product is hand-tight, rated to 6,000psi, has .2uL total swept volume, is versatile for any in-line filtration application, prevents damage to downstream components, incorporates Free-Turn™ architecture, and you can change filter elements without disconnecting from the system.
Catalog No. 50-113-271