Learn More
Optimize Technologies OPTI-SOLV In-Line Wrench-Tight Stainless Steel Holder
SDP
Supplier:  Optimize Technologies 100403686
Encompass_Preferred
The user-friendly Wrench-Tight OPTI-SOLV™ In-Line Filter system is well suited for a number of applications, such as high temperature, high-pressure environments where quick changes are imperative. The unique swivel design allows frits to be changed without removing the connecting tubing. Just plumb it once and leave it. The frictional forces generated at the piston/seal interface inevitably lead to particulate shedding. Placement of a filtering element between the pump and injector/ auto sampler removes these particulates, preventing problems with downstream components and costly downtime. Key Features include: an easy wrench-tight product that is rated to 6,000psi and has .2uL total swept volume. It comes packaged with fittings, is versatile for any in-line filtration application, prevents damage to downstream components, incorporates Free-Turn™ architecture, and you can change filter elements without disconnecting from the system.
Catalog No. 50-200-6152