Learn More
Optimize Technologies OPTI-SOLV Direct Connect Mini Filter, 0.5µm, 5pk
SDP
Supplier:  Optimize Technologies 100400095
Encompass_Preferred
The OPTI-SOLV™ Mini Filter provides low-impact filtration in a package no larger than a finger-tight fitting. Use the Mini Filter to extend the life of analytical columns or before mass spectrometers as a last line of defense against debris. Perfect zero-dead-volume connections are obtained every time with the patented automatic tube stop depth adjustment. Unique depth-filtering elements allow for greater capacity with no band spreading or loss of performance. The efficient design requires no holder and threads directly into any 10-32 port. It is available in 5 packs and recommended for volume-critical applications. For use up to 6,000psi.
Catalog No. 50-113-260