Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Puritan™ Opti- Tranz™ Agar Gel Media Transport Tube with Miniature Rayon Tip Swab

Catalog No. 22027133
Change view
Click to view available options
Swab Configuration:
Dual Applicator
Single Applicator
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Swab Configuration
22-027-133 Dual Applicator
22-027-132 Single Applicator
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. 22-027-133 Supplier Puritan™ Supplier No. ACL62PR

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Ideal for direct microscopic examination of clinical samples

4ml Amies Gel - without charcoal (clear)Aerobic and Anaerobic Culture. Ideal for direct microscopic examination of clinical samples. Double, standard rayon swabs on plastic handles. Rayon is non-toxic and made from USP grade fiber. Throat, vaginal, wound and skin swab sampling. Shelf Life: 24 Months. Made in USA

Specifications

Product Type Transport Tube
Media Type Semi-solid (Sodium Thioglycollate and Agar)
Sterility Sterile
Swab Configuration Dual Applicator
Closure Color Blue
Closure Type Screw Cap
Diameter (Metric) 5.15 mm
For Use With (Application) Throat, urogential, and wound swab sampling
Handle Material Polystyrene
Includes Charcoal additives
Tip Material Rayon
Tip Type Standard dual applicator
Tube Size 154 mm
Volume (Metric) 4 mL
Show More Show Less

For in vitro diagnostic (IVD) use only.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.