Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Puritan™ Opti- Tranz™ Agar Gel Media Transport Tube with Miniature Rayon Tip Swab

Catalog No. p-7111634
Click to view available options
Swab Configuration:
Dual Applicator
Single Applicator
This item is not returnable. View return policy
This item is not returnable. View return policy

Ideal for direct microscopic examination of clinical samples

4 mL Amies Gel - without charcoal (clear). Aerobic and Anaerobic Culture. Ideal for direct microscopic examination of clinical samples. Single, standard rayon tip with plastic handle. Rayon is non-toxic and made from USP grade fiber. Shelf Life: 24 Months. Made in USA

Specifications

Product Type Transport Tube
Media Type Semi-solid (Sodium Thioglycollate and Agar)
Sterility Sterile
Swab Configuration Single Applicator
Closure Color Blue
Closure Type Screw Cap
Diameter (Metric) 4.77 mm
For Use With (Application) Throat, urogential, and wound swab sampling
Handle Material Polystyrene
Includes Charcoal additives
Tip Material Rayon
Tip Type Standard single applicator
Tube Size 154 mm
Volume (Metric) 4 mL
Show More Show Less

For in vitro diagnostic (IVD) use only.

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.