Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Puritan™ Opti- Tranz™ Agar Gel Media Transport Tube with Miniature Rayon Tip Swab, Charcoal Additives

Catalog No. 22027139
Change view
Click to view available options
Shaft Material:
Aluminum
Polystyrene
Soft Aluminum
Closure Color:
Black
Orange
White
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Shaft Material Closure Color
22-027-139 Soft Aluminum Black
22-027-138 Aluminum Orange
22-027-136 Polystyrene Black
22-027-137 Polystyrene White
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Catalog No. 22-027-139 Supplier Puritan™ Supplier No. ACH61SAR

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

For preservation of aerobic, anaerobic, fastidious bacteria

4 ml Amies Gel w/ Charcoal. Aerobic and Anaerobic CultureFor aerobes, anerobes, and fastidious bacteria. Miniature tip helps minimize discomfort. Ear, nose, throat and eye sampling. Male ureathral sampling. Shelf Life: 24 Months. Made in USA

Specifications

Product Type Transport Tube
Media Type Semi-solid (Sodium Thioglycollate and Agar)
Sterility Sterile
Closure Color Black
Closure Type Screw Cap
Description Diameter: 5.15mm
Diameter (Metric) 5.15 mm
For Use With (Application) Preservation of samples
Includes Charcoal additives
Shaft Material Soft Aluminum
Tip Material Rayon
Tip Type Standard
Tube Size 154 mm
Volume (Metric) 4 mL
Show More Show Less

For in vitro diagnostic (IVD) use only.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.