Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Puritan™ Opti-Tranz™ Liquid Amies Media Transport Tube with Miniature Rayon Tip Swab

Catalog No. 22027120
Change view
Click to view available options
Tip Type:
Dual applicator
Minitip
Single applicator
Closure Color:
Orange
Red
White
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Tip Type Closure Color
22-027-120 Single applicator White
22-027-121 Dual applicator Red
22-027-122 Minitip Orange
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Catalog No. 22-027-120 Supplier Puritan™ Supplier No. LA61PR
This item is not returnable. View return policy

Ideal for direct aerobic culture, gram staining and rapid detection tests

1 mL Liquid Amies in absorbent polyurethane foam. Aerobic CultureIdeal for direct microscopic examination of clinical samples. Single, standard rayon swab with plastic handle. Throat, vaginal, wound and skin swab sampling. Shelf Life: 18 Months. Made in USA

Specifications

Product Type Transport Tube
Media Type Semi-solid (Sodium Thioglycollate and Agar)
Sterility Sterile
Swab Configuration Single Applicator
Closure Color White
Closure Type Screw Cap
Description Diameter: 5.15mm
Diameter (Metric) 5.15 mm
For Use With (Application) Throat, urogential, and wound swab sampling
Handle Material Polystyrene
Shaft Material Polystyrene
Tip Material Rayon
Tip Type Single applicator
Tube Size 154 mm
Volume (Metric) 1 mL
Show More Show Less

For in vitro diagnostic (IVD) use only.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.