Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Puritan™ Opti- Tranz™ Liquid Amies Media Transport Tube with Miniature Rayon Tip Swab, Aluminum Handle

Catalog No. 22027123
Change view
Click to view available options
Fill Volume:
1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Fill Volume
22-027-123 1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. 22-027-123 Supplier Puritan™ Supplier No. LA61SAR

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Used for isolation of aerobic culture

1 mL Liquid Amies in absorbent polyurethane foam. Aerobic CultureIdeal for direct microscopic examination of clinical samples. Rayon tips are non-toxic and made from USP grade fiber. Soft aluminum wire is flexible and can be easily shaped. Ear, nose, throat, and eye sampling, as well as male urethral sampling. Shelf Life: 18 Months. Made in USA

Specifications

Product Type Transport Tube
Media Type Semi-solid (Sodium Thioglycollate and Agar)
Sterility Sterile
Closure Color Green
Closure Type Screw Cap
Fill Volume 1 mL
For Use With (Application) Throat, urogential, and wound swab sampling
Shaft Material Soft Aluminum
Tip Material Rayon
Tip Type Minitip
Tube Size 154 mm
Show More Show Less

For in vitro diagnostic (IVD) use only.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.