Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Puritan™ Opti- Tranz™ Liquid Stuart Media Transport Tube with Miniature Rayon Tip Swab

Catalog No. 22027128
Change view
Click to view available options
Shaft Material:
Aluminum
Polystyrene
Soft Aluminum
Closure Color:
Green
Orange
Red
White
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Shaft Material Closure Color
22-027-128 Aluminum Orange
22-027-129 Soft Aluminum Green
22-027-126 Polystyrene Red
22-027-127 Polystyrene White
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Catalog No. 22-027-128 Supplier Puritan™ Supplier No. LS61AR
This item is not returnable. View return policy

Ideal for ear, nose, throat, male urethral sampling

1 ml Liquid Stuart in absorbent polyurethane foam, which acts as a moisture reservoir for bacteriaI. Intended for collection and transport of aerobic and fastidious bacteria. Used in bacterial culture, rapid tests, and Gram staining. Ear, nose, throat, and eye sampling, as well as male urethral sampling. Rayon tip is non-toxic and made from USP grade fiber. Miniature tip minimizes patient discomfort. Shelf Life: 18 Months. Made in USA

Specifications

Product Type Transport Tube
Media Type Semi-solid (Sodium Thioglycollate and Agar)
Sterility Sterile
Closure Color Orange
Closure Type Screw Cap
Description Diameter: 1.98mm
Diameter (Metric) 1.98 mm
For Use With (Application) Throat, urogential, and wound swab sampling
Shaft Material Aluminum
Tip Material Rayon
Tip Type Minitip
Tube Size 154 mm
Volume (Metric) 1 mL
Show More Show Less

For in vitro diagnostic (IVD) use only.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.