Learn More
Puritan™ Opti- Tranz™ Plus Liquid Amies Media Transport Tube with HydraFlock™ Tip Swab
Catalog No. 22027125
Change view
Click to view available options
Tip Type:
Dual applicator
Single applicator
Closure Color:
Pink
Yellow
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Tip Type Closure Color
22-027-125 Dual applicator Pink
22-027-124 Single applicator Yellow
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. 22-027-125 Supplier Puritan™ Supplier No. LA62H
Item Discontinued This item has been discontinued by the supplier. Please to view product availability in your area.
This item is not returnable. View return policy
Used for isolation of aerobic culture
1 ml Liquid Amies in absorbent polyurethane foam, which acts as a moisture reservoir for bacteria. Intended for collection and transport of aerobic and fastidious bacteria. Used in bacterial culture, rapid tests, and Gram staining. Flocked swab provides superior specimen collection and elution. Double HydraFlock™ swabs (3206-2H), plastic handles. Shelf Life: 18 Months. Made in USA
Specifications
Product Type Transport Tube
Media Type Semi-solid (Sodium Thioglycollate and Agar)
Sterility Sterile
Swab Configuration Dual Applicator
Closure Color Pink
Closure Type Screw Cap
For Use With (Application) Throat, urogential, and wound swab sampling
Handle Material Polystyrene
Tip Material Polyester Flock
Tip Type Dual applicator
Tube Size 154 mm
Volume (Metric) 1 mL
Show More Show Less
For in vitro diagnostic (IVD) use only.