Learn More
MilliporeSigma OPTI XLT 30 CAP MEDIA STERILE
Catalog No. NC3346479
Encompass
opticap XLT 30, sterile, inlet connection diam. 5/8 in., pore size 0.5/0.2 μm, cartridge nominal length 30 in. (75 cm)
Catalog No. NC3346479 Supplier MilliporeSigma Supplier No. KHGES3THH1
opticap XLT 30, sterile, inlet connection diam. 5/8 in., pore size 0.5/0.2 μm, cartridge nominal length 30 in. (75 cm)