Learn More
Glass Expansion Optic Fiber for D-Torch 170 mm
SDP
Catalog No. 502703149
Encompass_Preferred
Optic Fiber for D-Torch 170mm. Compatible with Thermo 6000 Duo ICP-OES. This shorter optical fiber assembly is for Thermo 6000 models up to the year 2010. From the year 2011 onwards, use Optic Fiber 450mm (31-808-3117).Glass Expansion is ISO 9001:2015 certified by AB Certification, No. AU4501-9001
Catalog No. 50-270-3149 Supplier Glass Expansion Supplier No. 318082933
Optic Fiber for D-Torch 170mm. Compatible with Thermo 6000 Duo ICP-OES. This shorter optical fiber assembly is for Thermo 6000 models up to the year 2010. From the year 2011 onwards, use Optic Fiber 450mm (31-808-3117).Glass Expansion is ISO 9001:2015 certified by AB Certification, No. AU4501-9001