Learn More
Glass Expansion Optic Fiber for D-Torch 450 mm
SDP
Catalog No. 502703145
Encompass_Preferred
Optic Fiber for D-Torch 450mm. Compatible with Thermo 6000 Duo ICP-OES. This longer optical fiber assembly is used from the year 2011 onwards. For Thermo 6000 models up to the year 2010, use the shorter optical fiber assembly (31-808-2933).Glass Expansion is ISO 9001:2015 certified by AB Certification, No. AU4501-9001
Catalog No. 50-270-3145 Supplier Glass Expansion Supplier No. 318083117
Optic Fiber for D-Torch 450mm. Compatible with Thermo 6000 Duo ICP-OES. This longer optical fiber assembly is used from the year 2011 onwards. For Thermo 6000 models up to the year 2010, use the shorter optical fiber assembly (31-808-2933).Glass Expansion is ISO 9001:2015 certified by AB Certification, No. AU4501-9001