Learn More
MSC SPI Brinell Bench Top Hardness Tester, Digital, 8 to 650 HBW, 1 ea
Catalog No. 5000512414
Encompass
The Brinell bench top hardness tester is an electronic instrument designed for precise material hardness measurements. This system provides reliable performance for evaluating various materials within an industrial testing environment.
  • Digital display for clear reading of results
  • Brinell scale measurement type
  • Hardness testing range of 8 to 650 HBW
  • Bench top configuration for stable operation
Catalog No. 50-005-12414 Supplier MSC Supplier No. 38443479
The Brinell bench top hardness tester is an electronic instrument designed for precise material hardness measurements. This system provides reliable performance for evaluating various materials within an industrial testing environment.
  • Digital display for clear reading of results
  • Brinell scale measurement type
  • Hardness testing range of 8 to 650 HBW
  • Bench top configuration for stable operation