Learn More
Bio Rad Laboratories OPTICAL FLAT 8-CAP STRIPS
Supplier:  Bio Rad Laboratories TCS0803
Encompass
Flat 8-cap strips tightly seal 0.2 ml tubes and plates for PCR and cold storage. They feature ultraclear upper surfaces, ideal for fluorescence or luminescence applications. Package contains 120 natural-color cap strips of 8. (TCS-0803)
Catalog No. 50-901-0032