Learn More
Thermo Scientific™ Optical-Quality Imaging Plates, 384-well, black-walled, clear bottom
Catalog No. A58941
Encompass_Preferred
Change view
Click to view available options
Quantity:
32 plates
Catalog No. Quantity
A58941 32 plates
1 options
Catalog No. A58941 Supplier Thermo Scientific™ Supplier No. A58941
  • $948.00
    $777.36 / Each of 1
    Save $170.64
    18% Off The following deviations and disclosures may produce prices below stated discounts but will not trigger price reductions: Price shown reflects a temporary, promotional price reduction. No other purchase or commercial obligation required to receive the reduced price. Price may change at any time. Promotional price valid on web orders only. Your contract pricing may differ. Other exclusions apply.
Add to Cart
Add to Cart
These imaging plates are optically transparent, extra-low, ultra-flat, 384-well plates that provide optimal performance for microscopy and plate-reader-based assays.
These imaging plates are optically transparent, extra-low, ultra-flat, 384-well plates that provide optimal performance for microscopy and plate-reader-based assays. The black-walled clear-bottom 384-well imaging plates come tissue-culture-treated, pre-sterilized, and labeled with unique barcodes. They are ideal for fluorescence high-content screening and imaging assays on our CellInsight High-Content Screening instruments, stand-alone microscopes such as the EVOS M7000/5000, and multimodal plate readers such as the Varioskan LUX microplate reader.
Features of these Imaging Plates include:
  • Quality results – no more out-of-focus cells. Optical-quality cyclo-olefin polymer (COP) provides best-in-class flatness (100 μm from edge to edge), transmittance, and glass-matching refractive index (1.34).
  • Extra-low base – skirt height of 0.21 mm allows for all wells and fields of view to ne read with immersion and high NA lenses
  • Faster read times – reduce autofocus search times due to the ultra-flat plate bottom. Improved transmittance of COP allows for shorter exposure times, which can reduce imaging times and minimize light exposure to the sample
  • Results you can trust – COP is ideal for fluorescence imaging assays since it is optically transparent in a broad wavelength range, along with being 100% biocompatible, chemically inert, and mechanically stable
  • Add and go – plates come tissue-culture treated and pre-sterilized, ready for immediate cell plating. Custom coatings available upon request
  • Screening ready – plates come with unique barcodes, ready for your high content screening assays. Custom barcodes available upon request
When running high-content screening and imaging assays, high image quality is essential for good results. Several factors can impair image quality, such as using thick, uneven plates, which can lead to focusing differences between wells and image quality issues across a plate. The optical properties of these imaging plates produce superior image quality and resolution from each well when used with fluorescence or luminescence readers. The flatness of the plate bottom also yields excellent performance in laser-based autofocus systems. Using these Optical-Quality Imaging Plates helps get the most from automated imaging systems and ensure that critical assays and samples are not compromised due to inferior plates.
TRUSTED_SUSTAINABILITY
Specifications
Barcode Alphanumeric
Lid With TSP Lid
Array 16 x 24
Autoclavable Not Autoclavable
Color Black
Disposable Single-use
Material Cyclic Olefin Polymer
No. of Wells 384
Sterility Sterile
Surface Treatment Tissue Culture
Well Shape Flat Bottom (F-well)
No. per Case 32 plates
Quantity 32 plates
Fit for Purpose Sterile and PCR Ready
Show More Show Less
For Research Use Only.