Learn More
Agilent Technologies OPTICAL STRIP CAPS 120/PK
Catalog No. NC9263434
Encompass
Optical Strip Caps, Mx3000P;, Strips of 8 Optically Clear Caps, to be Used With 96-Well Non-Skirted PCR Plate #410088
This item is not returnable. View return policy
Catalog No. NC9263434 Supplier Agilent Technologies Supplier No. 401425
Optical Strip Caps, Mx3000P;, Strips of 8 Optically Clear Caps, to be Used With 96-Well Non-Skirted PCR Plate #410088