Learn More
MilliporeSigma OPTICAP GAMMA XL300 3/PK
Supplier:  MilliporeSigma KHVEG003HH3
Encompass
Opticap Gamma Compatible XL 300 Millipore Express SHR with Prefilter 0.5/0.1 m 9/16 in. HB/HB 3/pk
Catalog No. NC1845906