Learn More
MilliporeSigma Opticap Sterile XL 10 Millipore Express SHC 0.5/0.2 µm 1-1/2 in. TC-9/16 in. HB
Supplier:  MilliporeSigma KHGES10TH1
Encompass
Opticap Sterile XL 10 Millipore Express SHC 0.5/0.2 µm 1-1/2 in. TC-9/16 in. HB
Catalog No. NC1813390