Learn More
MilliporeSigma Opticap XL150 Millipore Express SHC
Supplier:  MilliporeSigma KHGES015FB3
Encompass
Sterile, inlet connection diam. 3/4 in., pore size 0.5/0.2 μm, cartridge nominal length 3.8 in. (9.7 cm).
Catalog No. NC1271417