Learn More
MilliporeSigma Opticap Gamma Compatible XL 150 Millipore Express SHC 0.5/0.2 um 9/16 in. HB/HB
Supplier:  MilliporeSigma KHGEG015HH1
Encompass
Polypropylene (PP) capsule, 14 mm (9/16 in.) Hose Barb, 1/4 in. Hose Barb with double O-ring seal
Catalog No. NC1879577