Learn More
EMD MILLIPORE 3P BIOPHARM OPTICAP XL03 EXPRESS SHC GC 0.
Supplier:  EMD MILLIPORE 3P BIOPHARM KHGEG03HH3
Encompass
Opticap® Gamma Compatible XL 3 Millipore Express® SHC 0.5/0.2 µm 9/16 in. HB/HB3 Double Easy-Open bag
Catalog No. NC2133466