Learn More
MilliporeSigma OPTICAP XL10 DURAPORE
Supplier:  MilliporeSigma KHGLA10TH1
Encompass
Opticap XL10 Durapore sterile, inlet connection diam. 1.5 in., pore size 0.45/0.22 m, cartridge nominal length 10 in. (25 cm)
Catalog No. NC2229465