Learn More
EMD MILLIPORE 3P BIOPHARM OPTICAP XL10 EXPRESSSHC STER.
Supplier:  EMD MILLIPORE 3P BIOPHARM KHGES10TT1
Encompass
Opticap Sterile XL10 Millipore Express SHC 0.5 + 0.2um 1-1/2in. TC/TC
Catalog No. NC9960220