Learn More
MilliporeSigma Opticap XL4 Polysep II, 3/pk
Supplier:  MilliporeSigma KGW3A04HH3
Encompass
Inlet connection diam. 9/16 in., pore size 1.0/0.2 μm, cartridge nominal length 4 in. (10 cm).
Catalog No. NC1490329