Learn More
Chemglass Life Sciences 24-Position Block for 15 x 45mm 1 Dram (4mL) Flat Bottom Vials
SDP
Catalog No. 50871941
Encompass_Preferred
24-Position Block for 15 x 45mm 1 Dram (4mL) Flat Bottom Vials. Vial and tube holders for use in centrifugal vacuum evaporators. Blocks are designed to not only maximize evaporation but also to minimize the weight of the block, thereby increasing sample volumes. Constructed of aluminum, each block is black hard coated for resistance to organic solvents and laser etched with the vial/tube size.
Catalog No. 50-871-941 Supplier Chemglass Life Sciences Supplier No. OP660005
24-Position Block for 15 x 45mm 1 Dram (4mL) Flat Bottom Vials. Vial and tube holders for use in centrifugal vacuum evaporators. Blocks are designed to not only maximize evaporation but also to minimize the weight of the block, thereby increasing sample volumes. Constructed of aluminum, each block is black hard coated for resistance to organic solvents and laser etched with the vial/tube size.