Learn More
Chemglass Life Sciences 12-Position Block for 25 x 100mm Round Bottom Tubes, Two Tier Design
SDP
Catalog No. 50874148
Encompass_Preferred
12-Position Block for 25 x 100mm Round Bottom Tubes, Two Tier Design. Vial and tube holders for use in centrifugal vacuum evaporators. Blocks are designed to not only maximize evaporation but also to minimize the weight of the block, thereby increasing sample volumes. Constructed of aluminum, each block is black hard coated for resistance to organic solvents and laser etched with the vial/tube size.
Catalog No. 50-874-148 Supplier Chemglass Life Sciences Supplier No. OP660042
12-Position Block for 25 x 100mm Round Bottom Tubes, Two Tier Design. Vial and tube holders for use in centrifugal vacuum evaporators. Blocks are designed to not only maximize evaporation but also to minimize the weight of the block, thereby increasing sample volumes. Constructed of aluminum, each block is black hard coated for resistance to organic solvents and laser etched with the vial/tube size.