Learn More
Chemglass Life Sciences Optichem Block, 96-Position, 8 x 30mm Shell Vial Inserts
SDP
Catalog No. 507513689
Encompass_Preferred
Optichem Block, 96-Position, 8 x 30mm Shell Vial Inserts. Vial and tube holders for use in centrifugal vacuum evaporators. Blocks are designed to not only maximize evaporation but also to minimize the weight of the block, thereby increasing sample volumes. Constructed of aluminum, each block is black hard coated for resistance to organic solvents and laser etched with the vial/tube size.
Catalog No. 50-751-3689 Supplier Chemglass Life Sciences Supplier No. OP660004
Optichem Block, 96-Position, 8 x 30mm Shell Vial Inserts. Vial and tube holders for use in centrifugal vacuum evaporators. Blocks are designed to not only maximize evaporation but also to minimize the weight of the block, thereby increasing sample volumes. Constructed of aluminum, each block is black hard coated for resistance to organic solvents and laser etched with the vial/tube size.