Learn More
Chemglass Life Sciences 48-Position Block for 12 x 32mm HPLC Flat Bottom Vials
SDP
Catalog No. 50871940
Encompass_Preferred
48-Position Block for 12 x 32mm HPLC Flat Bottom Vials. Vial and tube holders for use in centrifugal vacuum evaporators. Blocks are designed to not only maximize evaporation but also to minimize the weight of the block, thereby increasing sample volumes. Constructed of aluminum, each block is black hard coated for resistance to organic solvents and laser etched with the vial/tube size.
Catalog No. 50-871-940 Supplier Chemglass Life Sciences Supplier No. OP660003
48-Position Block for 12 x 32mm HPLC Flat Bottom Vials. Vial and tube holders for use in centrifugal vacuum evaporators. Blocks are designed to not only maximize evaporation but also to minimize the weight of the block, thereby increasing sample volumes. Constructed of aluminum, each block is black hard coated for resistance to organic solvents and laser etched with the vial/tube size.