Learn More
Med Vet International Opticlar LED Ophthalmoscope Equipment - Ophthalmoscope Battery Set, J1662A, Each
Catalog No. 5000583901
Encompass_Preferred
JorVet has partnered with Opticlar, which is a UK based manufacturer of high-end LED diagnositic equipment such as Otoscopes and Opthalmoscopes. LED lighting has many benefits with better colors and most importantly lasts a long time. These units come with a 10-year bulb warranty! The Opticlar LED Opthalmoscopes takes veterinary diagnostic instruments to a new level. We spoke to veterinary practices at length to understand exactly what they wanted ? and the Vision-Vet range has been designed and manufactured with all the features and benefits that Veterinarians have long been demanding, providing superb diagnostics and competitive pricing throughout. Contents: ? Opthalmoscope head w/LED light ? ?C? cell battery handle ? Zip case ? 5-year warranty
Catalog No. 50-005-83901 Supplier Med Vet International Supplier No. JOR1662A
JorVet has partnered with Opticlar, which is a UK based manufacturer of high-end LED diagnositic equipment such as Otoscopes and Opthalmoscopes. LED lighting has many benefits with better colors and most importantly lasts a long time. These units come with a 10-year bulb warranty! The Opticlar LED Opthalmoscopes takes veterinary diagnostic instruments to a new level. We spoke to veterinary practices at length to understand exactly what they wanted ? and the Vision-Vet range has been designed and manufactured with all the features and benefits that Veterinarians have long been demanding, providing superb diagnostics and competitive pricing throughout. Contents: ? Opthalmoscope head w/LED light ? ?C? cell battery handle ? Zip case ? 5-year warranty