Learn More
StatLab OpticMount X Mounting Medium
SDP
Catalog No. 21033253
Change view
Click to view available options
Volume (Metric):
118 mL
Catalog No. Volume (Metric)
21-033-253 118 mL
1 options
Catalog No. 21-033-253 Supplier StatLab Supplier No. 7722
Add to Cart
Add to Cart
Xylene based solution, ensure proper adhesion and transparency of the tissue for clear microscopic observation
  • Used to permanently attach a tissue section to a microscope slide by fixing it under a coverslip, essentially mounting the specimen for examination
  • Formulation utilizes xylene as a solvent to ensure proper adhesion and transparency of the tissue for clear microscopic observation
  • 4 oz.
Specifications
Product Type Mounting Medium
Reagent Type Mounting Solution
Volume (Metric) 118 mL