Learn More
Vernier™ Optics Expansion Kit
SDPSCI_ED
Catalog No. S16054ND Shop All Vernier Software Products
Change view
Click to view available options
Includes:
Light Source, three lenses, screen and holder, power supply
Catalog No. Includes
S16054ND Light Source, three lenses, screen and holder, power supply
1 options
Catalog No. S16054ND Supplier Vernier™ Supplier No. OEK
Add to Cart
Add to Cart
Includes
Combination LED point and luminous light source with power supply, Three lenses integrated with holders (100, 200, and —150 mm focal length), Screen and holder, Light Sensor holder, Aperture screen and holder, Power supply
Add the Vernier Optics Expansion Kit to your Dynamics Cart and Track System to conduct optics experiments, such as image formation with lenses and light intensity vs. distance. You can even use the kit to build a basic telescope.
  • Combination LED point and luminous light source with power supply
  • Three lenses integrated with holders (100, 200, and 150 mm focal length)
  • Screen and holder
  • Light Sensor holder
  • Aperture screen and holder
  • Power supply
  • Add the Vernier Optics Expansion Kit to your Dynamics Cart and Track System to conduct optics experiments, such as image formation with lenses and light intensity vs. distance. You can even use the kit to build a basic telescope.
  • The parts of the Optics Expansion Kit snap on to the track, so that the parts can be left in place while the track is moved. The three lenses are permanently mounted in three plastic supports. The light source is LED-based, so that it uses less energy and is cool to the touch. A rotating disc on the light source gives you the option of a radiant object, a point source, an L with 2 by 1 cm dimension, and parallel lines for resolution experiments. The power supply is the same as the LabQuest power supply. An additional aperture plate is included for experiments based on lens diameter and shape.
Warranty and Services
5-year limited warranty
Specifications
For Use With (Application) Educational use only
Includes Light Source, three lenses, screen and holder, power supply
WARNING: Cancer and Reproductive Harm - www.P65Warnings.ca.gov