Learn More
Cell Systems Corporation OptiCulture™ Serum-Free Medium-RH (4E3-500-RH)
SDP
Catalog No. NC3957530
Encompass_Preferred
OptiCulture™ Serum-Free Medium-RH (4E3-500-RH) is a xeno-free, nutrient-rich medium designed to support reproducible growth and functional phenotypes across diverse cell types. Eliminating all non-human animal-derived components, this medium provides a clean, defined environment for research requiring animal-free conditions. The kit includes 500 mL basal medium, OptiBoom-R (recombinant human growth factors), supplements (ITS, Anti-Oxidants, Hydrocortisone), and 10 mL xeno-free coating material. 4E3-500-RH is certified for experimental use, phenol-red free, and contains no added hormones or antibiotics. Ideal for xeno-free barrier models, organoids, co-cultures, disease modeling, and drug screening.
Catalog No. NC3957530 Supplier Cell Systems Corporation Supplier No. 4E3500RH
OptiCulture™ Serum-Free Medium-RH (4E3-500-RH) is a xeno-free, nutrient-rich medium designed to support reproducible growth and functional phenotypes across diverse cell types. Eliminating all non-human animal-derived components, this medium provides a clean, defined environment for research requiring animal-free conditions. The kit includes 500 mL basal medium, OptiBoom-R (recombinant human growth factors), supplements (ITS, Anti-Oxidants, Hydrocortisone), and 10 mL xeno-free coating material. 4E3-500-RH is certified for experimental use, phenol-red free, and contains no added hormones or antibiotics. Ideal for xeno-free barrier models, organoids, co-cultures, disease modeling, and drug screening.