Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

GeminiBio Optima Fetal Bovine Serum
SDP

Catalog No. 11852144 Shop All Gemini Bio Products
Change view
Click to view available options
Grade:
Optima
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Grade
11-852-144 Optima
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 11-852-144 Supplier GeminiBio Supplier No. S12450
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

High-quality, cost-effective serum with high lot-to-lot consistency for reproducible general cell culture.

  • High-quality, cost-effective FBS for general cell culture applications
  • 100% US-origin material for traceability and compliance
  • Low endotoxin and hemoglobin for excellent cell growth
  • Sterility-, virus-, and performance-tested with a full biochemical profile
  • Triple 0.1 μm sterile-filtered
  • High lot-to-lot consistency for reproducible research
  • Ships on dry ice; store frozen at -20°C to -10°C
  • Formerly sold under the Atlanta Biologicals and Bio-Techne/R&D Systems brands
  • Complimentary heat inactivation upon request

Specifications

Additional Assays Biochemical profile
Certifications/Compliance ISO 13485:2016 certified quality system
Content And Storage 500 mL bottle, store frozen at -20°C to -10°C
Species Bovine
Age Fetal
Country of Origin United States
Form Liquid
Format Bottle
Product Type Fetal Bovine Serum
Quantity 500 mL
Serum Treatment Standard (Sterile-filtered)
Sterility Sterile
Sterilization Method Triple-filtered, 0.1 μm
Source Fetal bovine blood
Grade Optima
Processing Triple 0.1 μm sterile-filtered
Tested For Mycoplasma, Sterility (bacteria and fungi), Adventitious viruses per 9 CFR 113.53 (BVDV, IBRV, PI-3V, BTV, BRSV, BPV, BAV-3, BAV-5, Reovirus, Rabies)
Endotoxin Concentration ≤20.0 EU/mL
Formulation Liquid, frozen
For Use With (Application) Cell Culture
Heat Inactivated No
Hemoglobin Concentration ≤25 mg/dL
Lyophilized No
Product Line Optima
Mycoplasma Detected No
Osmolality 280 to 335 mOsm/kg
pH 6.8 to 7.8
Shipping Condition Dry Ice
Test Methods LAL assay (endotoxin); USP methods (sterility); 9 CFR 113.53 (virus)
USDA Safety Testing Included Yes
Regulatory Status RUO
Gamma-irradiated No
Show More Show Less

For cell culture, further manufacturing or research use only. Not for direct therapeutic use.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.