Learn More
SCP Science Hydrogen peroxide, PPP, 30%, 500ml
Catalog No. NC0965191
Encompass
PlasmaPURE Plus acids and reagents are manufactured with trace metal impurity levels less than 10 ppt (0.01ppb) for ultra low level analysis below 1ppb. Supplied in preleached Teflon bottles for optimum quality and packaged in Class 10 clean room conditions. Complete with a Certificate of Analysis with lot number, expiry date and maximum specification for over 60 analytes. Volume discounts available.
Catalog No. NC0965191 Supplier SCP Science Supplier No. 250036145
PlasmaPURE Plus acids and reagents are manufactured with trace metal impurity levels less than 10 ppt (0.01ppb) for ultra low level analysis below 1ppb. Supplied in preleached Teflon bottles for optimum quality and packaged in Class 10 clean room conditions. Complete with a Certificate of Analysis with lot number, expiry date and maximum specification for over 60 analytes. Volume discounts available.