Learn More
Bellco Glass, Inc. Optima Holding Tray,10.5 SQ. For Plastic Bag
SDP
Catalog No. 501945971
Encompass_Preferred
Holding tray for plastic bags. Manufactured from clear high quality acrylic for rockers and shakers. Temperatures up to 50 degrees Celsius. The optima holding tray is designed to fit many types of laboratory shakers and rocking platforms. Place holding tray on top of a platform that has a rubber mat. MADE IN THE USA
Catalog No. 50-194-5971 Supplier Bellco Glass, Inc. Supplier No. 764432000
Holding tray for plastic bags. Manufactured from clear high quality acrylic for rockers and shakers. Temperatures up to 50 degrees Celsius. The optima holding tray is designed to fit many types of laboratory shakers and rocking platforms. Place holding tray on top of a platform that has a rubber mat. MADE IN THE USA