Learn More
Beckman Coulter Beckman Coulter Optima MAX-TL Ultracentrifuge, tabletop, 120,000 rpm, 657,000 x g
Catalog No. NC2816536
Encompass
The Beckman Coulter Optima MAX-TL ultracentrifuge is a compact tabletop instrument designed for high-speed separations up to 120,000 rpm and 657,000 x g. It provides a glove-friendly color LCD touchscreen, multilingual software, and multi-level biosafety features for routine laboratory ultracentrifugation.
  • Tabletop ultracentrifuge with maximum speed of 120,000 rpm.
  • Maximum relative centrifugal force 657,000 x g.
  • Compatible with TL-series fixed-angle rotors and common labware.
  • Color LCD touchscreen with glove-friendly interface.
  • Multilingual software available in nine languages.
  • Multi-level biosafety options and optional pharmaceutical-grade sterilizing filter.
  • Designed to fit in standard biosafety hoods.
Catalog No. NC2816536 Supplier Beckman Coulter Supplier No. A95761
To request a quote for this item, please select Request a Quote to reach our dedicated support team.
The Beckman Coulter Optima MAX-TL ultracentrifuge is a compact tabletop instrument designed for high-speed separations up to 120,000 rpm and 657,000 x g. It provides a glove-friendly color LCD touchscreen, multilingual software, and multi-level biosafety features for routine laboratory ultracentrifugation.
  • Tabletop ultracentrifuge with maximum speed of 120,000 rpm.
  • Maximum relative centrifugal force 657,000 x g.
  • Compatible with TL-series fixed-angle rotors and common labware.
  • Color LCD touchscreen with glove-friendly interface.
  • Multilingual software available in nine languages.
  • Multi-level biosafety options and optional pharmaceutical-grade sterilizing filter.
  • Designed to fit in standard biosafety hoods.