Learn More
GRANT INSTRUMENTS Heating circulator digital, general purpose -25°C* to 120°C, pump
SDP
Catalog No. NC2165563
Encompass_Preferred
Optima TC120 Heated Circulating Bath.The Grant TC120 series of digital heating circulating baths for general applications requiring precision temperature control in the range of -15°C to 120°C.The TC120 heating circulator can be used in combination with any Grant stainless steel tanks or with a T-Clamp for independent use of virtually any vertical sided tank.Cooling/heating range -15°C to 120°C.Stability ±0.05ºC / Uniformity ±0.1ºC.Tank options 5L to 38L.Simple, intuitive user interface: dial and two function buttons.3 programmable temperature presets.Clear, bright 4 digit LED display.Low level protection and adjustable over temperature cut outCountdown timer with audible alarm.
This item is not returnable. View return policy
Catalog No. NC2165563 Supplier GRANT INSTRUMENTS Supplier No. TC120 US
Optima TC120 Heated Circulating Bath.The Grant TC120 series of digital heating circulating baths for general applications requiring precision temperature control in the range of -15°C to 120°C.The TC120 heating circulator can be used in combination with any Grant stainless steel tanks or with a T-Clamp for independent use of virtually any vertical sided tank.Cooling/heating range -15°C to 120°C.Stability ±0.05ºC / Uniformity ±0.1ºC.Tank options 5L to 38L.Simple, intuitive user interface: dial and two function buttons.3 programmable temperature presets.Clear, bright 4 digit LED display.Low level protection and adjustable over temperature cut outCountdown timer with audible alarm.